Specifications
- Cat. No.:76028-732
- Max. operating temperature:37.8 °C (100 °F)
Specifications
About this item
3M™ High Flow Series Dual Flow Twin Manifold Assembly (DF2XX) provide a vehicle for appropriate water quality to equipment requiring different water characteristics for cold beverages, ice and coffee. Manifolds are innovatively designed for ease of installation and maintenance. This model includes a pressure gauge to monitor differential water pressure.
- ¾" Inlet/Outlet for optimal water flow in cold beverages, ice and coffee
- Optional features include expansion heads for increased capacity (select High Flow Series models only)
- Multiple manifold design options for specialized application needs
- Pressure gauge allows easy system pressure monitoring (excludes SH–EXP)
They are made for multiple pieces of equipment to address various water filtration conditions. Assemblies lend themselves to versatility and are ideal for constructing customized filtration systems, for multiple equipment applications, that are effective and help save time and additional labor costs.
Ease of installation and maintenance. This model includes a pressure gauge to monitor differential water pressure.