3M™ NanoSHIELD™ Hollow Fiber Series Filter Cartridge
NanoSHIELD™ hollow fiber series PP filter cartridges have been engineered to combine the highest level of particle retention with superior flow rates and minimal pressure drop for the most demanding applications in the latest technology node.
- Hollow Fiber Technology provides up to 2 times more surface area as compared to equivalent sized pleated filters, and increased depth (2x thicker) of filtration results in improved particle retention
- Large Surface Area for up to 5 times higher flow rates as compared to other pleated cartridges, 10" NanoSHIELD hollow fiber series cartridges flow similar to 20" pleated cartridges for increased lifetime, throughput, and overall equipment effectiveness (OEE)
- Compact Design allows for smaller, less costly filter housings and reduces hold-up volume, exposure, and waste of expensive chemicals
- 0.005 to 0.1 μm retention ratings offer superior reduction of particles, gels, and micro-bubbles, as well as reduced micro-bridge and wafer level defects
- Quality Manufactured in a cleanroom to reduce particle adders and extractables
- Filters are critically cleaned and tested for process repeatability
- The NanoSHIELD hollow fiber series family of products is offered in several configurations to allow the end user to match the volume of fluid with the appropriate filter in an effort to reduce chemical waste
The hollow fiber PP membrane in an all PE construction provides low metallic/ionic contamination and excellent resistance to most chemicals.
End configuration: 222 O-Ring and Flat Cap
Frequently Bought Together