About this item
Zeta Plus™ S Series Filter Cartridges to provide optimal clarification of food and beverage, cosmetic and industrial fluids. The cartridges are constructed with Zeta Plus™ S Series depth filter media, which is composed of inorganic filter aid, cellulose, and a resin system that imparts a positive charge to the filter matrix, aiding its adsorption of negatively-charged contaminants.
- Used in a variety of applications in the food and beverage and cosmetic industries
- Exhibit high contaminant-holding capacity and provide economical filtration and reliable particle reduction
- Retain contaminants by mechanical entrapment and electrokinetic adsorption
- Superior particle reduction
Offered in a full range of scalable capsule and cartridge filter configurations, enabling pilot testing and scale-up operations to use the same material as in full-scale systems.
Our family of Zeta Plus™ S Series Filter Cartridges are composed of advanced cellulose-based depth filtration media, designed to retain contaminants by both mechanical entrapment and electrokinetic adsorption. Zeta Plus S Series depth filter media is composed of high surface area filter aids embedded in a cellulose fiber matrix. During the manufacturing process, the resin system incorporated into the filter media imparts a positive charge to the filter matrix to permanently form a filtration structure with positively charged electrokinetic capture sites. The resulting porous depth filter is a tortuous network of charge-enhanced flow channels capable of removing bacteria, particulate, and submicronic contaminants to a level which mechanical screening alone cannot achieve.
Our cartridges are constructed from individual cells of Zeta Plus™ S Series filter media, assembled together with PP ring seals under predetermined compression, and unitized by three, 316 stainless steel bands. Each cell is constructed using polypropylene molded edge-seals and separators for high performance. Gasket material options are available.
Applications: Beverages, wine, beer, juices, soft drinks, cider, distilled spirits, liquor, cognac, cordials, food products, syrups, brines, broths, vinegar, edible oils, sugars, cosmetics, perfumes, lotions, shampoos, oils, deodorants and colognes.
We also offer a range of cartridge sizes, available in 8", 12" and 16" diameters, with surface areas ranging from 0.26 to 3.9 m²(2.8 to 42.2 ft²) per cartridge to make scale-up straightforward and predictable.