3M™ CUNO Legacy Large Diameter Drop-In Style Replacement Cartridge, Model CFS210-2, 5618907
Supplier: 3M Healthcare
Certificates
About this item
3M™ CUNO legacy large diameter drop–in style replacement cartridge, Model CFS210–2, when used in a drop–in style filtration housing, provides consistent quality water by reducing particulate.
- 20" large diameter drop–in style replacement cartridge
- Provides consistent quality water
- Reduces particulate from incoming water
- For use in CFS22 or CFS22–1.5 drop–in style filtration housings
Certifications: *As tested and verified by manufacturer's laboratory.
Frequently Bought Together