ContactUsLinkComponent
3M™ OCS In-line System for Office Coffee Applications, Model CS-341, 5560216
  76030-071
3M™ OCS In-line System for Office Coffee Applications, Model CS-341, 5560216
  76030-071
 :  7100051445
  • Cat. No.:
    76030-071
  • Model:
    5560216
3M™ Water Filtration Products OCS In-line Systems for Office Coffee Applications provide you with consistent quality water for hot beverages that can help protect your equipment and elevate profitability. Our water filtration in-line systems are an inexpensive way to help protect your equipment from potentially harmful scale build-up that can decrease production and profitability.
  • Provides consistent quality water for coffee brewers (including single-serve) and hot water dispensers by reducing sediment*, chlorine taste and odor and scale*
  • Reduces the ability of calcium and magnesium to form as hard scale* on the dispenser nozzle and heating coils which can reduce downtime and unscheduled service maintenance
  • Reduction of sediment* helps prevent equipment wear
  • Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change-outs
  • Compact design allows for installation in tight spaces
3M™ Water Filtration Products OCS In-line systems for Office Coffee Applications are designed to be effective at reducing sediment*, chlorine taste and odor, and scale* allowing you to feel more confident in the quality of the products you serve and providing protection for your equipment
In addition, the Sanitary Quick Change (SQC) design allows for fast and easy cartridge change-outs that are sanitary by design – requiring no contact with the filter media during cartridge change-out.
3M™ Water Filtration Products branded replacement cartridges and filter systems offer a wide range of solutions designed to address many types of water quality concerns. Third-party certifications, such as NSF and WQA, are an advantage provided on many 3M Water Filtration Products. 3M Purification is a known leader in filtration technology recognized for innovation, trust and quality around the world.
*Tested and verified by manufacturer’s laboratory.