ContactUsLinkComponent
3M™ OCS System for Office Coffee Applications featuring Rogo Filter Head Design, Model CS-451, 5588111
3M™ OCS System for Office Coffee Applications featuring Rogo Filter Head Design, Model CS-451, 5588111
Catalog # 76030-229
Supplier:  3M Healthcare
3M™ OCS System for Office Coffee Applications featuring Rogo Filter Head Design, Model CS-451, 5588111
Catalog # 76030-229
Supplier:  3M Healthcare
Supplier Number:  7100051041
Specifications
  • Cat. No.:
    76030-229
  • Model:
    5588111
Specifications
About this item
3M™ Water Filtration Products OCS System for Office Coffee Applications featuring Rogo Filter Head Design provides you with consistent quality water for hot beverages that can help increase repeat business and help to protect your equipment from hard scale build-up.
  • Provides consistent quality water for office coffee and hot cup applications by reducing sediment* and chlorine taste and odor
  • Reduces the ability of calcium and magnesium to form as hard scale* helping to reduce downtime and unscheduled service maintenance
  • Reduction of sediment* can help prevent equipment wear
  • Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change-outs
3M™ Water Filtration Products OCS System for Office Coffee Applications featuring Rogo Filter Head Design for brewed coffee, tea and hot chocolate is designed to be effective at reducing sediment*, chlorine taste and odor, and scale* allowing you to feel more confident in the quality of the products you serve and can provide equipment protection from scale build-up
The Sanitary Quick Change (SQC) design allows for fast and easy cartridge change-outs that are sanitary by design - requiring no contact with the filter media during cartridge change-out.
3M™ Water Filtration Products replacement cartridges and filter systems offer a wide range of solutions designed to address many types of water quality concerns. Third-party certifications, such as NSF and WQA, are an advantage provided on many 3M Water Filtration Products. 3M Purification is a known leader in filtration technology recognized for innovation, trust and quality around the world.
*Tested and verified by manufacturer's laboratory.