ContactUsLinkComponent
3M™ OCS Water System for Espresso Machines & Tea Brewers, Model VH3/OWS120-L, 5609243
3M™ OCS Water System for Espresso Machines & Tea Brewers, Model VH3/OWS120-L, 5609243
Catalog # 76030-285
Supplier:  3M Healthcare
3M™ OCS Water System for Espresso Machines & Tea Brewers, Model VH3/OWS120-L, 5609243
Catalog # 76030-285
Supplier:  3M Healthcare
Supplier Number:  7100049912
Specifications
  • Cat. No.:
    76030-285
  • Model:
    5609243
Specifications
About this item
3M™ Water Filtration Products OCS Water System for Espresso Machines & Tea Brewers featuring Cyst, Lead and Asbestos Reduction provides consistent high quality water by reducing sediment*, cyst**, chlorine taste and odor, lead and asbestos from incoming water. The system’s internal flow control contributes to the lead reduction capabilities by allowing more contact time with media.
  • Provides consistent high quality water for espresso machines and tea brewers by reducing sediment*, cyst**, chlorine taste and odor, lead and asbestos from incoming water
  • No external shut-off valve required due to built-in valve-in-head design
  • Head with internal flow control to allow more contact time with media, reducing lead that may be in the water
  • Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change-outs without the need for a filter wrench
3M™ Water Filtration Products OCS Water System for Espresso Machines & Tea Brewers featuring Cyst, Lead and Asbestos Reduction is designed to be effective at reducing sediment*, cyst**, chlorine taste and odor, lead and asbestos, allowing you to feel more confident with the quality of your water.
With no external shut-off valve required due to the built-in valve-in-head design and the head with internal flow control allowing more contact time with media, this filtration system can be a great choice for your needs.
3M™ Water Filtration Products systems and cartridges offer a wide range of solutions designed to address many types of water quality concerns. Third-party certifications, such as NSF and WQA, are an advantage provided on many 3M Water Filtration Products. 3M Purification is a known leader in filtration technology recognized for innovation, trust, and quality around the world.
*As tested and verified by manufacturer's laboratory.
**Based on the use of Cryptosporidium parvum oocysts.