3M™ CUNO Legacy Drop-In Style Replacement Cartridge for Ice Maker Applications, Model CFS420IMF, 5560905
Supplier: 3M Healthcare
Certificates
About this item
3M™ CUNO legacy drop–in style replacement cartridge for ice maker applications, Model CFS420IMF, when used in a drop–in style filtration housing, provides consistent quality water by reducing particulate. The built–in scale inhibitor helps to reduce potential harmful effects of scale build–up on equipment.
- 9³/₄" drop–in style replacement cartridge
- Provides consistent quality water
- Reduces particulate from incoming water
- Graded density design built–in scale inhibitor helps to reduce potential harmful effects of scale build–up on equipment
Frequently Bought Together