3M™ CUNO Legacy In-line System for Beverage Applications, Model CFS717, 5560215
Catalog # 76030-069
Supplier: 3M Healthcare
Specifications
- Cat. No.:76030-069
- Model:5560215
Specifications
About this item
3M™ CUNO legacy in–line system for beverage applications, Model CFS717 provides consistent quality water by reducing particulate and chlorine taste and odor. Built–in scale inhibition can help to prevent the potentially harmful effects of scale build–up on equipment.
- Provides consistent quality water
- Reduces particulate and chlorine taste and odor from incoming water
- Built–in scale inhibition can help to prevent the potentially harmful effects of scale build – up on equipment
- ¼" NPT inlet and outlet allows for easy connections to existing piping
- Compact in – line design provides convenient point–of–use filtration in tight space applications
- System designed for use in coffee and hot tea applications