ContactUsLinkComponent
3M™ DC Series Filter Housing Part
3M™ DC Series Filter Housing Part
Catalog # 76029-358
Supplier:  Avantor
3M™ DC Series Filter Housing Part
Catalog # 76029-358
Supplier:  Avantor
Specifications
  • Cat. No.:
    76029-358
Specifications
About this item
The 3M™ DC series filter housing is constructed of durable 304L stainless steel, meeting general purpose industrial and commercial filtration needs. All wetted welded surfaces are stainless steel and external surfaces of these filter housings are electro-polished for an easy-care finish. The filter housings are available in a variety of sizes to accommodate from 4 to 88 filter cartridges.
  • 304L stainless steel construction provides excellent corrosion protection
  • Accepts 9¾ or 10" modular cartridges, in 1-, 2-, 3-, or 4- high configurations for flexibility in equipment sizing
  • V-Band clamp and swing bolt designs optimize cartridge change-out
  • Removable cartridge posts for ease in cleaning
3M™ DC series filter housing models 12 DC and 22 DC include spring and seal assemblies that permit the use of filter cartridges with single length equivalents of 9¾ or 10". The 4 DC and 5 DC models use pressure plate assemblies, while the 12 DC and 22 DC models use pressure plate with individual spring and seal assemblies. Standard DC housings are configured to accept double open-end style cartridges. To adapt the housing to accept single open end, double o-ring cartridges with locating spear, remove the housing cartridge posts and the spring and seal assembly. Replace the standard pressure plate and the center rod with the single open end (SOE) pressure plate and center rod (sold separately).
Applications: chemicals and petrochemicals; food and beverage; industrial processes; paint and coatings; industrial water treatment.