ContactUsLinkComponent
3M™ Bypass Plug, TFS450, 5623902
  76031-680
3M™ Bypass Plug, TFS450, 5623902
  76031-680
 :  7000051197
  • Cat. No.:
    76031-680
Bypass Plugs are a range of plugs to fit 3M™ filter heads as appropriate.
  • Enables water to flow directly through the filter head
  • Use for taking out of use one of the filter heads in a manifold system
  • Use when 'wintering' a system
  • Come in a range of sizes
3M™ Bypass Plugs to enable water to flow directly through the filter head when replacing the filter cartridge. Our plugs can be used for replacing out one of the heads in a manifold system, and for 'wintering' a system. They come in a range of sizes to fit 3M heads as appropriate.
Taking one of the heads in a manifold system out of use, the plugs enable water to flow directly through the head by replacing the filter cartridge.