ContactUsLinkComponent
3M™ Commercial Reverse Osmosis Water Storage Tanks, Model 20 Gal. Drawdown Tank w/Connection Kit, 5598408
3M™ Commercial Reverse Osmosis Water Storage Tanks, Model 20 Gal. Drawdown Tank w/Connection Kit, 5598408
Catalog # 76030-244
Supplier:  3M Healthcare
undefined
3M™ Commercial Reverse Osmosis Water Storage Tanks, Model 20 Gal. Drawdown Tank w/Connection Kit, 5598408
Catalog # 76030-244
Supplier:  3M Healthcare
Supplier Number:  7000029227
Specifications
  • Environmentally Preferable:
  • Cat. No.:
    76030-244
  • Filter type:
    Tank
Specifications
About this item
3M™ commercial reverse osmosis water storage tanks, model 20 gal. Drawdown tank with connection kit, 5598408 is appropriately sized for boiler-based steamer units up to 300000 BTUs.
  • Reverse osmosis 20 gallon drawdown tank at 60 psi with 15 psi pre-charge
  • Polyethylene tank liner and fiberglass wrap tank exterior
Drawdown tank with connection kit, 5598408 contains a polyethylene tank liner and fiberglass wrap tank exterior. Tanks can be plumbed together to accommodate larger steamer operations. Fittings are designed to accept plumbing from 3M™ reverse osmosis systems.