Order Entry
United States
ContactUsLinkComponent
3M™ Water Filtration System, Model CFS8576-S, 5581906
3M™ Water Filtration System, Model CFS8576-S, 5581906
Catalog # 76030-217
Supplier:  3M Healthcare
3M™ Water Filtration System, Model CFS8576-S, 5581906
Catalog # 76030-217
Supplier:  3M Healthcare
Supplier Number:  7000050279

Specifications

  • Cat. No.:
    76030-217
  • Model:
    5581906

Specifications

About this item

Water filtration system designed for hot beverage and ice applications.

  • Capacity: 10000 gal. (37850 L) at 1.5 gpm (5.6 lpm)

One micron sediment filtration, scale inhibition, chlorine taste and odor reduction.