Specifications
- Micron Rating:2.5 Absolute
- Cat. No.:76031-763
- Filter type:Depth filter
- Model:1GPK1DP09025
- Max. operating temperature:71 °C (160 °F)
- Filter material:PP
Specifications
About this item
CUNO™ CTG-Klean system filter pack with Betafine™ XL series filter cartridge is a totally enclosed filtration system, using a separate pressure vessel and disposable filter pack to isolate the product from the housing. Cartridge pack design reduces time and labor involved with change-outs and cleaning of the housing, helping to reduce worker exposure to cleaning solvents.
- Totally enclosed filter pack avoids handling of used filter cartridges
- Design reduces gaskets, posts, spring and seal assemblies, and seal plates
- Filter pack can be capped and reused, depending on application
- Rapid filter change-out reduces labor and the need for duplex systems
- Easy to use design minimizes down time
- Consistent, absolute-rated, pleated PP media provides more surface area in less physical space and a longer service life than non-pleated filters
A self-contained pack containing Betafine XL series filter cartridges is installed into a special housing.
The product being filtered is contained within the filter pack and does not come in contact with the housing. Our factory sealed filter packs prevent by-pass and help reduce exposure of workers to hazardous fluids. The self-contained system simplifies the filter change-out process, and reduces both cleaning and downtime. Our streamlined system, coupled with high performance, long-life filters, make CUNO CTG-Klean System Filter Packs the clear choice for efficient and cost-effective filtration. In some applications, hold up volume can be recovered from the system and used filter packs can be capped and stored for repeated use, lowering costs. The Betafine™ XL Series Filter difference.
These filters are used extensively on corrosive and non-corrosive process fluids where broad chemical compatibility is required. Our line of Betafine™ XL series filter cartridges are ideally suited for high flow, low viscosity fluids. The vast surface area of the pleated filter matrix offers increased flow, high contaminant loading capacity and low initial pressure drop. Betafine XL series filter construction is optimized to obtain high flow rates while maintaining the defined particle reduction efficiencies. Each filter cartridge is manufactured from 100% PP and pleated for increased surface area.
Made to Filter.
The Power of Pleated Performance
Our precision engineered, absolute rated, pleated filter provides dependable, repeatable filtration, improving effluent quality and helps provide consistent performance. The nature of pleated media also constitutes a greater surface area in less physical space and provides a longer service life compared to non-pleated media. Greater surface area leads to fewer filter change-outs and lower total filtration operating costs. The greater surface area means higher flow rates with lower pressure drops, which yields exceptional throughput, reduced processing time and reduced processing costs.
Our thermally bonded, 100% PP construction affords extensive chemical and thermal compatibility and no media migration.
Versatility
Our Betafine™ XL series filters provide superior chemical resistance and durability in demanding process applications such as:
Coatings - Maintain high quality in feed streams and intermediates, Electronics - RO/DI pre-filtration, electroplating baths, process/ rinse water, solvent, specialty coatings, Food and Beverage- Potable water, process and blending water and diatomaceous earth trap filtration in food and beverage applications and ndustrial Process- Filtration of intermediates, fine chemicals, and photographic chemicals, reagent grade chemicals, high purity chemicals, oil and gas processing, secondary water filtration, and process gases.
Superior on-line service life provides significant total filtration cost reductions. From fewer filter cartridges used to a reduction in labor costs by decreasing filter change-out frequency, Betafine XL series filters give you the ultimate in cost effective pleated filter technology.
Applications include Resins, varnishes, glues, Photographic solutions, Fine chemicals, Special coatings, Process and waste water, Food and beverage, Lubricating and cutting oils, High quality chemicals, Contaminated water, Ceramic slurries, CMP slurries and Hazardous chemicals.