To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Species:HIV
- Size:1 mg
- Storage Conditions:–20 °C
- Protein Synonyms:TAT Peptide linker GGG|TAT (47-57) GGG|TAT (47-57)|TAT Cys(Npys) GGG linker
- Protein/Peptide Name:TAT (47-57) GGG-Cys(Npys)
- Purity:95%
- Molecular Weight:1987.3 Da
- Sequence:YGRKKRRQRRRGGG-C(Npys)-NH2
- Cat. No.:103006-428
- Supplier no.:AS-61215
This peptide corresponds to the protein transduction domain of the TAT protein and is synthesized with an activated cysteine residue C(Npys), wherein Npys is 3-Nitro-2-pyridinesulfenyl group and is used for activating S of cysteine and for rapid reaction when a thiol group is introduced. The N-terminus is rendered free for applications requiring certain conjugation reactions with a free N-terminal end, and a linker GGG is placed at the C-terminal end, and the peptide has been synthesized with the C(Npys) group. This kind of modification has been used to render this peptide as a cell penetrating and carrier peptide applicable in conjugation studies.
Sequence: YGRKKRRQRRRGGG-C(Npys)-NH2
MW: 1987.3 Da
% Peak area by HPLC: 95
Storage condition: -20°C