Order Entry
United States
ContactUsLinkComponent
 
Human Angiotensin II Substrate
Catalog # 103003-754
Supplier:  Anaspec Inc
CAS Number:  
Human Angiotensin II Substrate
Catalog # 103003-754
Supplier:  Anaspec Inc
Supplier Number:  AS-24520
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Human
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Ang Peptide chymosin substrate|Ang II|Ang II substrate|Chymosin substrate Peptide|Angiotensin II substrate|human Ang II substrate
  • Protein/Peptide Name:
    Angiotensin II Substrate
  • Purity:
    95%
  • Molecular Weight:
    1126.2 Da
  • Sequence:
    DRV-pY-IHPF
  • Cat. No.:
    103003-754
  • Supplier no.:
    AS-24520

Specifications

About this item

This is a tyrosine phosphorylated angiotensin II peptide with a substrate sequence specificity for chymase to act. This is an octapeptide hormone that is formed as a result of the action of human heart chymase, a chymotrypsin-like serine protease on the Phe-His bond of Angiotensin I. The ideal susbtrate for human heart chymase should contain this peptide sequence, and has been demonstrated that the Pro-Phe sequence in P2-P1 positions of peptide substrates is highly favored by leukocyte chymotrypsin-like proteinases such as chymases and cathepsin G.
Sequence: DRV-pY-IHPF
MW: 1126.2 Da
% Peak area by HPLC: 95
Storage condition: -20°C