Order Entry
United States
ContactUsLinkComponent
Human Myelin oligodendrocyte glycoprotein
Human Myelin oligodendrocyte glycoprotein
Catalog # 103007-806
Supplier:  Anaspec Inc
CAS Number:  
Human Myelin oligodendrocyte glycoprotein
Catalog # 103007-806
Supplier:  Anaspec Inc
Supplier Number:  AS-63918
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Human
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Myelin oligodendrocyte|Myelin oligodendrocyte (35-55)|MOG
  • Protein/Peptide Name:
    Myelin oligodendrocyte glycoprotein
  • Purity:
    95%
  • Molecular Weight:
    2592 Da
  • Sequence:
    MEVGWYRPPFSRVVHLYRNGK
  • Cat. No.:
    103007-806
  • Supplier no.:
    AS-63918

Specifications

About this item

Myelin oligodendrocyte glycoprotein (MOG) is a member of the immunoglobulin superfamily and is expressed exclusively in the central nervous system. It is a glycoprotein observed to be important in the myelination of nerves. It is a central molecule actively studied for its role in Multiple Sclerosis. MOG (35-55) is able to induce autoantibody production and relapsing-remitting neurological disease causing extensive plaque-like demyelination. Autoantibody response to MOG (35-55) has been observed in multiple sclerosis (MS) patients and MOG (35-55)-induced experimental autoimmune encephalomyelitis (EAE) C57/BL6 mice and Lewis rats.
Sequence: MEVGWYRPPFSRVVHLYRNGK
MW: 2592 Da
% Peak Area by HPLC: ≥ 95%
Storage condition: -20°C