Rapid Fixer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
A rapid-acting hardening fixer for automated and tray processing
- Complete fixer
- Kit makes 1 gallon
- 32 oz solution A and 3.6 oz solution B
Complete fixer.