Order Entry
United States
ContactUsLinkComponent
SEM Large Double Slot Vise, Electron Microscopy Sciences
SEM Large Double Slot Vise, Electron Microscopy Sciences
  103302-508
SEM Large Double Slot Vise, Electron Microscopy Sciences
  103302-508
 :  75951-04
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Description:
    Large Double Slot Set Screw Vise
  • Size:
    25 mm
  • Environmentally Preferable:
  • Cat. No.:
    103302-508
  • Supplier no.:
    75951-04

 

 

Clamp thin specimens and cross sections without conductive paint or tape for viewing in a scanning electron microscope using the double slot vise specimen holder.

  • Use for paper, metals, plastic, textiles, plant material and more
  • Machined aluminum with stainless steel allen set screws

2.5 mm wide x 5 mm deep slots
Dimensions: 25 x 8 mm (1 x 0.32") with 3.2 mm (1/8") pin

 : Includes allen wrench.