Order Entry
United States
ContactUsLinkComponent
Uranium (U) Pure Single-Element Standard, 1000 µg/ml, HO, 125 ml, PerkinElmer
Uranium (U) Pure Single-Element Standard, 1000 µg/ml, HO, 125 ml, PerkinElmer
Catalog # 101687-668
Supplier:  Avantor
CAS Number:  
undefined
Uranium (U) Pure Single-Element Standard, 1000 µg/ml, HO, 125 ml, PerkinElmer
Catalog # 101687-668
Supplier:  Avantor
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    H₂O
  • Size:
    125 ml
  • Cat. No.:
    101687-668
  • Element:
    Uranium
  • Supplier no.:
    N9303844

Specifications

About this item

Uranium (U) Pure Grade Atomic Spectroscopy Calibration Standard is supplied with a comprehensive Certificate of Analysis that documents the quality and reliability. Matrix: HO Volume: 125 mL.

  • GHS-Signal words - US-Warning
  • GHS-Symbols - US-GHS07