Tiger Paw Coated Abrasive Flap Discs, Weiler®
Supplier: Safety & Industrial Supplies
Certificates
About this item
Multiple layers of coated abrasive material expose sharp, fresh grains as they slowly wear away, and the 100% premium zirconium grain prevents premature grain shedding on aggressive applications resulting in increased productivity and reduced operating costs.
- Designed for aggressive performance on tough grinding applications
- High performance grain and special poly cotton cloth provide superior grain retention
- This combination gets the job done faster and reduces overall grinding and finishing costs
Frequently Bought Together