ContactUsLinkComponent
Proto® Midget Angle Nose Plier, ORS Nasco
Proto® Midget Angle Nose Plier, ORS Nasco
Catalog # 300024-172
undefined
Proto® Midget Angle Nose Plier, ORS Nasco
Catalog # 300024-172
Supplier Number:  577-235
Specifications
  • Length:
    4¹⁄₁₆"
  • Cat. No.:
    300024-172
  • Jaw width:
    ⅝"
  • Jaw length:
    ¾"
  • Supplier no.:
    577-235
  • Jaw thickness:
    ¹¹⁄₆₄"
Specifications
About this item
Precision machined mating surfaces. Plastisol grips for added comfort.