Order Entry
United States
ContactUsLinkComponent
Instrument Calibration Standard 2, Matrix per Volume 5% HNO₃, PerkinElmer
Instrument Calibration Standard 2, Matrix per Volume 5% HNO₃, PerkinElmer
Catalog # 101674-702
Supplier:  Avantor
CAS Number:  
undefined
Instrument Calibration Standard 2, Matrix per Volume 5% HNO₃, PerkinElmer
Catalog # 101674-702
Supplier:  Avantor
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Instrument Calibration Standard 2 (5% HNO₃), 125 mL, 400 mg/L: Ni, 200 mg/L: Zn, 150 mg/L: Mn, 100 mg/L: Ag, Cr
  • Cat. No.:
    101674-702
  • Supplier no.:
    N9300219

Specifications

About this item

Instrument Calibration Standard 2, Matrix per Volume 5% HNO₃ per 100 ml, Concentration of Element: 100 mg/ml Ag,100 mg/ml Cr, 150 mg/ml Mn, 400 mg/ml Ni, and 200 mg/ml Zn. We recommend a 100-fold dilution with high-quality 5% HNO₃. All the solutions you need for daily calibration of your ICP and HGA tor CLP analyses. Remember to prepare your calibration standards fresh for each analysis and discard after use, Hazard Symbol: C.

  • Risk Phrases-R34
  • Safety Phrases-S17-20-23-26-29-36/37/39-45-60
  • H phrases US-H314-H317-H351
  • P phrases US-P260u-P280g-P280i-P261-P280-P264-P272-P201-P202-P303+P361+P353-P305+P351+P338-P310a-P304+P340-P363-P308+P313-P321-P333+P313-P301+P330+P331-P302+P352a-P405-P501a
  • GHS-Signal words - US-Danger
  • GHS-Symbols - US-GHS05 GHS08 GHS07