Pipet-Lite™ XLS+™ Single Channel Manual Pipettes, Rainin, Mettler-Toledo
Supplier: METTLER TOLEDO RAININ LLS MS
Certificates
About this item
Pipet-Lite™ XLS+™ blends cutting-edge engineering and innovation to deliver highly reproducible results with exceptional comfort. The elastomeric seals and polymer tip ejectors, along with LTS™ LiteTouch System™, deliver smoothness and control.
- Ergonomic handle design with comfortable fingerhook
- Improved volume adjustment lock prevents drift
- Built-in shock absorber inside the tip ejector reduces thumb impact
- Autoclavable shaft assembly
- Corrosion-free, easy to clean tip ejector
The sure-fit handle, light springs, and 'Magnetic Assist™' technology ensure smooth, light operation and significantly reduce the risk of repetitive strain injuries.
The embedded RFID tag enables advanced calibration tracking with optional reader, software, and PC.
Frequently Bought Together