To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:Glass bottle
- Description:T7.Tag
- Size:1 kit
- Environmentally Preferable:
- Storage Temperature:+2 °C to +8 °C
- Cat. No.:EM69025-3
- Supplier no.:69025-3
Specifications
About this item
T7.Tag Affinity Purification Kit is designed for rapid immunoaffinity purification of target proteins that carry the T7.Tag sequence ( the 11 amino terminal aa of the T7 gene 10 protein).
- pH: 2.2
- Multiple toxicity
Purification is based on binding target proteins to T7.Tag monoclonal Aantibody that is covalently coupled to cross-linked agarose beads and can wash away unbound proteins. Neutralization Buffer is included to limit protein exposure to low pH. The beads can be used in either batch or column methods and can be regenerated a minimum of five times without loss of binding activity.