Order Entry
United States
Orders LinkContactUsLinkComponent
Wizard™ pH Buffer Crystallization Screen, Rigaku

Wizard™ pH Buffer Crystallization Screen, Rigaku

Wizard™ pH Buffer Block, 96 x 1 mL
Catalog # 101444-060
Marketsource
Supplier:   Rigaku Reagents
Supplier #:  1008654
Wizard™ pH Buffer Crystallization Screen, Rigaku
Catalog # 101444-060
Supplier:  Rigaku Reagents
Marketsource
Wizard™ pH Buffer Crystallization Screen, Rigaku
Wizard™ pH Buffer Crystallization Screen, Rigaku

Wizard™ pH Buffer Block, 96 x 1 mL

Price
$379.23
Log in for availability
Each (1ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

The Wizard™ pH Buffer screen is an optimization screen that can be applied to a crystallization hit optimization to simultaneously explore the effects of pH and buffer composition on crystal growth.

The Wizard™ pH Buffer screen is a set of 96 unique buffer solutions encompassing 12 unique buffer systems at 8 pH points, incrementally varied by 0.4 pH units. The 96-well block plate formulations are 1.0mL each in a 96 deep-well matrix block plate. All formulations are prepared with ASTM type 1 water and high-purity chemicals, 0.2µm filtered in sterile packaging.

Documents

Specifications
  • Description:
    Wizard™ pH Buffer Block
  • Size:
    96 x 1 mL

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.