About this item
The PEG Ion series matrix combines the crystallizing power of polyethylene glycols (PEGs) with salts that are commonly found in crystallization conditions
Use PEG Ion during the coarse screening phase of biological macromolecules (proteins, nucleic acids, peptides, and combinations thereof).
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Related Information
Wizard PEG Ion 48 salt screen with PEG 1K and 4K Wizard PEG Ion 48 salt screen with PEG 8K and 10K