To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Brand Name:CAYM
- Cat. No.:75816-762
- Contents:Kit contains: Buffer, Standard, Reagent, Mixture, and Plate
- Range:2 - 20 mg/L
- Supplier no.:10010755-96
Simple, reproducible, and sensitive tool for measurement of glycerol in plasma and serum.
- Assay 40 samples in duplicate
- Plate-based colorimetric measurement (530 – 550 nm)
Glycerol is the backbone of triglycerides and an important intermediate in energy metabolism being involved in both oxidative and synthetic processes. The circulating levels of glycerol and free fatty acids are considered to reflect lipolysis and are therefore useful parameters to evaluate in various conditions. The measurement of glycerol is also useful for correction of glycerol interference when measuring triglycerides in reference materials and in serum from patients with elevated glycerol concentrations. The assay employs a coupled enzymatic reaction system that yields a brilliant purple product with an absorbance maximum at 540 nm.