Heavy Duty Racks for Microtubes, Globe Scientific
Supplier: Globe Scientific
Certificates
About this item
This popular, rugged, reinforced polypropylene rack provides strength and durability.
- Autoclavable
- Rubber feet provide stability on slick surfaces
- Numeric markings for sample identification
- Compatible with both 1.5 and 2.0 ml microtubes
With rubber feet providing stability, these racks can be used as a workstation on the benchtop.
Frequently Bought Together