2-Propanol solution ≥90% (v/v)
Supplier: Aqua Solutions
Certificates
About this item
CAS Number:67-63-0
:
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674
:60.1 g/mol
UN:1219
ADR:3,II
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Related Information
Certificates
Search by Lot Numberghs_I7613
SDS
spec_Isopropyl Alcohol 90% Solution
Datasheet
spec_Isopropyl Alcohol 90% vv Solution
Product specification
Frequently Bought Together