ContactUsLinkComponent
iworx® Small Animal ECG / EMG Systems
iworx® Small Animal ECG / EMG Systems
Supplier:  IWORX SYSTEMS INC.
Certificates
undefined
iworx® Small Animal ECG / EMG Systems
Supplier:  IWORX SYSTEMS INC.
Specifications
  • Catalog No:
  • 470231-654
  • 470231-656
  • Measurement Time:
  • 2000 samples/s per channel simultaneous
  • 2000 samples/s per channel simultaneous
  • Interface:
  • USB
  • USB
  • Software:
  • LabScribe
  • LabScribe