Carboxymethyl cellulose sodium salt, 800 - 3100 mPas (2%, H₂O, 25°C), white solid, high viscosity, Millipore®
Catalog # 80502-036
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:250 g
- Cat. No.:80502-036
- Supplier no.:217274-250GM
Specifications
About this item
:
Storage Temperature:Ambient
CAS Number:9004-32-4
Formula:[C₆H₇O₂(OH)x(OCH₂COONa)y]n
MDL Number:MFCD00081472