ContactUsLinkComponent
SmartCompliance Triangular Bandage Refill, First Aid Only
SmartCompliance Triangular Bandage Refill, First Aid Only
Catalog # 76049-114
Supplier:  First Aid Only
undefined
SmartCompliance Triangular Bandage Refill, First Aid Only
Catalog # 76049-114
Supplier:  First Aid Only
Supplier Number:  FAE-5100-001
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    SmartCompliance Triangular Bandage Refill
  • Environmentally Preferable:
  • Cat. No.:
    76049-114
Specifications
About this item
Relieve tension and support arm or shoulder injuries with this all-purpose triangular bandage.
  • Latex-free
  • Supplied in a sterile package
  • Reduces pressure on arms, elbows, or shoulders
  • Two safety pins included
  • Meets ANSI/ISEA Z308.1-2003 standards
Bandages are dispensed from the Smart Tab, ezRefill box, which includes an indicator tab to alert users when to reorder. The box snaps easily into place in all SmartCompliance cabinets, keeping supplies organized. For use with SmartCompliance systems: 90732, 90829, 90830, 90831, 90832, 90833, 90834, 90835.