ContactUsLinkComponent
Stretch-Knit Spray Sock Hoods, Allegro®
Stretch-Knit Spray Sock Hoods, Allegro®
Catalog # 300006-112
undefined
Stretch-Knit Spray Sock Hoods, Allegro®
Catalog # 300006-112
Supplier Number:  037-1410-12
Specifications
  • Clothing Size:
    One size
  • Type:
    Stretch-knit spray sock hoods
  • Garment application:
    Industrial,
    Paint and lacquer
  • Gender:
    Unisex
  • Fabric:
    Knit
  • Cat. No.:
    300006-112
  • Supplier no.:
    037-1410-12
Specifications
About this item
Provides comfortable protection against overspray and other particulates
Spray Sock fits over the head and can be worn with goggles, half mask, or full mask respirators.