Flame Resistant Coverall, ANSI Class 3 Compliant, Tecasafe Plus, Lakeland Industries
Supplier: Lakeland
Certificates
About this item
Tecasafe Plus Flame Resistant Coverall for primary protection from flash fire and electrical arc flash, where High Visibility Compliance is needed.
- Inherently FR Fabric that can't lose its flame resistant properties
- NFPA 2112, UL Certified for Flash Fire Protection
- NFPA 70E, CAT2 Electrical Arc Flash Protection
- ANSI 107-Class 3 Compliant for Hi-Vis
- Pleated back and elastic waist for fit and comfort
- Zippered leg cuffs for ease of donning and doffing
Tecasafe Plus High-Visibility Flame Resistant Coverall, dual certified for both flash fire and electrical arc flash protection, as well as ANSI 107-Class 3 for reflectivity. Designed for fit and comfort with pleated back, elastic waist, and plenty of pockets. Flared leg openings make donning and doffing easier, and sleeve cuffs feature adjustable snap closures.
Certifications: NFPA 2112. NFPA 70E, CAT2. ANSI 107-Class 3.
Specifications
- Catalog No:
- 75923-464
- 75923-462
- 75923-478
- 75923-476
- 75923-474
- 75923-472
- 75923-470
- 75923-468
- 75923-466
- Garment application:
- Industrial, Fire protection, Laboratory
- Industrial, Fire protection, Laboratory
- Industrial, Fire protection, Laboratory
- Industrial, Fire protection, Laboratory
- Industrial, Fire protection, Laboratory
- Industrial, Fire protection, Laboratory
- Industrial, Fire protection, Laboratory
- Industrial, Fire protection, Laboratory
- Industrial, Fire protection, Laboratory
- Risk area:
- HiVis, Flame-retardant
- HiVis, Flame-retardant
- HiVis, Flame-retardant
- HiVis, Flame-retardant
- HiVis, Flame-retardant
- HiVis, Flame-retardant
- HiVis, Flame-retardant
- HiVis, Flame-retardant
- HiVis, Flame-retardant
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening