7 oz. 88/12 Flame Resistant Coverall, Khaki, Lakeland Industries
: Lakeland
Flame Resistant Coverall for primary protection from flash fire and electrical arc flash.
- Lightweight 7 oz. 88/12 Flame Resistant fabric
- NFPA 2112, UL Certified for Flash Fire Protection
- NFPA 70E, CAT2 Electrical Arc Flash Protection
- Pleated back and elastic waist for fit and comfort
- Flared leg cuffs for ease of donning and doffing
Lakeland 7 oz. 88/12 Flame Resistant Coverall, dual certified for both flash fire and electrical arc flash protection. Designed for fit and comfort with pleated back, elastic waist, and plenty of pockets. Flared leg openings make donning and doffing easier, and sleeve cuffs feature adjustable snap closures.
: NFPA 2112. NFPA 70E, CAT2.
- Catalog No:
- 75923-646
- 75923-648
- 75923-650
- 75923-652
- 75923-654
- 75923-502
- 75923-656
- 75923-644
- Garment application:
- Industrial, Fire protection, Laboratory
- Industrial, Fire protection, Laboratory
- Industrial, Fire protection, Laboratory
- Industrial, Fire protection, Laboratory
- Industrial, Fire protection, Laboratory
- Industrial, Fire protection, Laboratory
- Industrial, Fire protection, Laboratory
- Industrial, Fire protection, Laboratory
- Risk area:
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- Fabric:
- Pre-shrunk 88/12 FR cotton
- Pre-shrunk 88/12 FR cotton
- Pre-shrunk 88/12 FR cotton
- Pre-shrunk 88/12 FR cotton
- Pre-shrunk 88/12 FR cotton
- Pre-shrunk 88/12 FR cotton
- Pre-shrunk 88/12 FR cotton
- Pre-shrunk 88/12 FR cotton
- Weight:
- 7 oz.
- 7 oz.
- 7 oz.
- 7 oz.
- 7 oz.
- 7 oz.
- 7 oz.
- 7 oz.
Frequently Bought Together