Orders LinkContactUsLinkComponent
Proto® Puller Step Plates, ORS Nasco
Proto® Puller Step Plates, ORS Nasco
Proto® Puller Step Plates, ORS Nasco
Certificates
Proto® Puller Step Plates, ORS Nasco
Proto® Puller Step Plates, ORS Nasco
13 Options Available Below
Product Details & Documents
Provides forcing screw base for hollow shaft applications 3/4 to 23/8" bore.
Product Family Options
Product Information
  • Diameter
    Weight
    Availability
    Price
  • 300024-950
    Puller Step Plate
    Puller Step Plate
    3/4 – 1"
    0.06 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $11.59
    $11.59
    Each
       $11.59
    Minimum order quantity is 1
    Supplier Number: 577-4040-1
  • 300024-962
    Puller Step Plate
    Puller Step Plate
    7/8 – 11/8"
    0.08 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $13.97
    $13.97
    Each
       $13.97
    Minimum order quantity is 1
    Supplier Number: 577-4040-2
  • 300024-964
    Puller Step Plate
    Puller Step Plate
    1 – 11/4"
    0.11 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $14.88
    $14.88
    Each
       $14.88
    Minimum order quantity is 1
    Supplier Number: 577-4040-3
  • 300024-966
    Puller Step Plate
    Puller Step Plate
    11/8 – 13/8"
    0.13 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $16.53
    $16.53
    Each
       $16.53
    Minimum order quantity is 1
    Supplier Number: 577-4040-4
  • 300024-968
    Puller Step Plate
    Puller Step Plate
    11/4 – 15/8"
    0.19 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $17.49
    $17.49
    Each
       $17.49
    Minimum order quantity is 1
    Supplier Number: 577-4040-5
  • 300024-970
    Puller Step Plate
    Puller Step Plate
    13/8 – 13/4"
    0.23 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $20.03
    $20.03
    Each
       $20.03
    Minimum order quantity is 1
    Supplier Number: 577-4040-6
  • 300024-972
    Puller Step Plate
    Puller Step Plate
    11/2 – 17/8"
    0.29 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $24.58
    $24.58
    Each
       $24.58
    Minimum order quantity is 1
    Supplier Number: 577-4040-7
  • 300024-974
    Puller Step Plate
    Puller Step Plate
    15/8 – 2"
    0.33 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $25.81
    $25.81
    Each
       $25.81
    Minimum order quantity is 1
    Supplier Number: 577-4040-8
  • 300024-976
    Puller Step Plate
    Puller Step Plate
    13/4 – 21/8"
    0.4 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $28.80
    $28.80
    Each
       $28.80
    Minimum order quantity is 1
    Supplier Number: 577-4040-9
  • 300024-952
    Puller Step Plate
    Puller Step Plate
    17/8 – 23/8"
    0.49 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $37.77
    $37.77
    Each
       $37.77
    Minimum order quantity is 1
    Supplier Number: 577-4040-10
  • 300024-954
    Puller Step Plate
    Puller Step Plate
    2 – 21/2"
    0.59 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $40.40
    $40.40
    Each
       $40.40
    Minimum order quantity is 1
    Supplier Number: 577-4040-11
  • 300024-956
    Puller Step Plate
    Puller Step Plate
    21/8 – 25/8"
    0.65 lbs.
    Discontinued
    Supplier Number: 577-4040-12
  • 300024-958
    Puller Step Plate
    Puller Step Plate
    21/4 – 23/4"
    0.76 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $45.37
    $45.37
    Each
       $45.37
    Minimum order quantity is 1
    Supplier Number: 577-4040-13
  • 300024-960
    Puller Step Plate
    Puller Step Plate
    23/8 – 27/8"
    0.84 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $49.48
    $49.48
    Each
       $49.48
    Minimum order quantity is 1
    Supplier Number: 577-4040-14