ContactUsLinkComponent
Xenon Lamps for Fluorescence Detectors, PerkinElmer
Xenon Lamps for Fluorescence Detectors, PerkinElmer
Catalog # 97019-248
Supplier:  PerkinElmer
undefined
Xenon Lamps for Fluorescence Detectors, PerkinElmer
Catalog # 97019-248
Supplier:  PerkinElmer
Supplier Number:  N2922082
Specifications
  • Description:
    Xenon Lamp for Series 200 and 200a Fluorescence Detector
  • Cat. No.:
    97019-248
  • Supplier no.:
    N2922082
Specifications
About this item
The Xenon source (150 W) used in Series 200 Fluorescence Detector provides signal to noise ratio of greater than 700:1 from trace analysis.