Order Entry
United States
Orders LinkContactUsLinkComponent
Test Tubes, Caplugs

Test Tubes, Caplugs

16 ml, PP, 17 mm, 100 mm
Catalog # 80700-708
Supplier:   Caplugs
Supplier #:  214-2380-030
Test Tubes, Caplugs
Catalog # 80700-708
Supplier:  Caplugs
Test Tubes, Caplugs

16 ml, PP, 17 mm, 100 mm

Price
$168.57
Log in for availability
Case of 1,000
Product Details & Documents

This polypropylene tube is more chemically resistant and are appropriate for use with organic solvents and stronger acids (except oxidizing acids), alkalis, alcohols, chlorinated & aromatic hydrocarbons (except xylene) and ketones.

Specifications
  • Ø:
    17 mm
  • Capacity:
    16 ml
  • Height:
    100 mm
  • Material:
    PP
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.