Topgard Replacement Suspensions, MSA
Catalog # 300081-036
Supplier: Safety & Industrial Supplies
Specifications
- Color:Gray
- Description:Staz-On® 4-Pt. Suspension, Size Standard; 61/2 - 8
- Material:Plastic
- Cat. No.:300081-036
- For use with:Topgard® Hat; Topgard® Cap
- Supplier no.:454-454232
Specifications
About this item
Designed for easy clip-in to the V-Gard® hat or cap shell
- Features Staz-On® suspension system
- 4-point suspension keeps helmet in place
- Liner can be adjusted for a more secure and comfortable fit
Includes sweatband and nylon height adjusters.