To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Environmentally Preferable:
- Detection Method:Confocal Imaging,Fluorescence,High Content Imaging
- Cat. No.:101676-436
- Storage temperature:19...25 °C
- Supplier no.:6005430
Our ViewPlate microplates are engineered for bottom reading systems and high content imaging
- Harmonized Code US-3926909910
- Material-Polystyrene, Glass Bottom
Glass exhibits better optical properties and is perfectly flat, but is less-suited for cell culture. Glass therefore usually has to be coated with proteins that enhance cell adherence and growth (e.g., poly-D-lysine or collagen). This high quality microplate can be utilized in your cell based assay for superior results. Leveraging years of assay and instrument experience in plate detection, PerkinElmer designs better microplates for better performance that guarantees better results for all PerkinElmer applications. Glass bottom microplates from PerkinElmer are not recommended to be used within an elevated temperature environment such as an incubator.