- Description:Escherichia coli derived from ATCC® BAA-2452™
- Environmentally Preferable:
- Cat. No.:76169-228
Everything needed to grow reference cultures for QC testing is included in this one handy device.
- Over 900 strains available
- All-in-one design reduces the risk of contamination
- Strains are less than or equal to four passages from reference culture
- Ready-to-use format saves time and money
- Refrigerated storage is easy and economical
- Online Certificate of Analysis provides detailed strain information
- Traceability to reference cultures ensures authenticity
- Technical Support experts available for guidance
KWIK-STIK’s simple, all-inclusive design makes life easier for lab techs while reducing chances for errors. Each KWIK-STIK contains a qualitative lyophilized microorganism pellet, ampoule of hydrating fluid, and inoculating swab. Available in a 2-pack format, universally used for all types of microbiological control testing.