Order Entry
United States
ContactUsLinkComponent
 
Staphylococcus aureus Recombinant Protein A (from E. coli), HiLyte Fluor® 750
Catalog # 103010-700
Supplier:  Anaspec Inc
CAS Number:  
Staphylococcus aureus Recombinant Protein A (from E. coli), HiLyte Fluor® 750
Catalog # 103010-700
Supplier:  Anaspec Inc
Supplier Number:  AS-72239
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    HiLyte Fluor® 750
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Staphylococcus aureus
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Gene ID:
    AAB05743.9
  • Protein/Peptide Name:
    Protein A
  • Purity:
    > 95% as determined by SDS-PAGE
  • Cat. No.:
    103010-700
  • Supplier no.:
    AS-72239

Specifications

About this item

Protein A-HiLyte™ Fluor 750 Conjugate can be used as a universal reagent to detect primary antibodies in Western Immunoblot from many species including rabbit, human, some mouse IgG isotypes, and others. Fluorescence (near-infrared) Excitation/Emission wavelength: 754 nm/778 nm.
AnaSpec's recombinant Protein A consists of only IgG binding domains and was expressed using a recombinant bacterial expression system. Its apparent molecular weight is approximately 30,000 Da compared to 42,000 Da for the native Protein A that is found in Staphylococcus aureus.
Protein A conjugates are widely used as labeled reagents for a variety of experiments including immunoprecipitation, antibody detection and purification and assay development