Order Entry
United States
ContactUsLinkComponent
Proto® Standard Length Round Head Ratchets, ORS Nasco
Proto® Standard Length Round Head Ratchets, ORS Nasco
Catalog # 300092-854
undefined
Proto® Standard Length Round Head Ratchets, ORS Nasco
Catalog # 300092-854
Supplier Number:  577-5452F

Specifications

  • Description:
    Standard Length Round Head Ratchet, 5° Angle
  • Length:
    9⅜"
  • Cat. No.:
    300092-854
  • Head type:
    Round Head
  • Finish:
    Full polish
  • Drive size:
    1/2"

Specifications

About this item

Small head design and tight swing allow fastener to access tight areas.

  • 5° ratcheting angle
  • 1.5" head width
  • Full polish shaft with knurled grip for strong slip-resistant grip
Certifications: ASME B107.10M.