ContactUsLinkComponent
Heavy-Duty Scratch Brushes with Scraper, Weiler®
Heavy-Duty Scratch Brushes with Scraper, Weiler®
Catalog # 300044-676
undefined
Heavy-Duty Scratch Brushes with Scraper, Weiler®
Catalog # 300044-676
Supplier Number:  804-44069
Specifications
  • Bristle material:
    Steel
  • Handle material:
    Wood
  • Total length:
    11 1/2"
  • Cat. No.:
    300044-676
  • No. of rows:
    4×11
  • Width:
    1½" block
  • Supplier no.:
    804-44069
  • Trim length:
    1½"
Specifications
About this item
For cleaning dirt, rust, scale, chips, and paint.
  • Sanded hardwood block finish
  • 0.0120" bristle diameter