ContactUsLinkComponent
No. 567 V-Block and Clamp, L.S. Starrett
Catalog # 300034-423
undefined
No. 567 V-Block and Clamp, L.S. Starrett
Catalog # 300034-423
Supplier Number:  681-52588
Specifications
  • Description:
    No. 567 V-Block and Clamp
  • Cat. No.:
    300034-423
  • Clamp extension:
    1⁵⁄₁₆"
Specifications
About this item
Can be used on its base, on the end, or on either side
The V at the stepped end is at right angles to the base and is handy for holding shouldered studs, pins, etc. A clearance hole for drilling or removing dowel pins is provided in the block. The block has four 3/8 - 16 tapped holes, and is hardened and precision ground. The clamp is a strong forging.
The clamp is smaller than the outside width of the block, but has an adjustable side screw to support the block and prevent tilting.