Proto® Long Hex Key Sets, Stanley
Catalog # 300002-900
Supplier: Safety & Industrial Supplies
Specifications
- Description:Long Hex Key Sets
- Cat. No.:300002-900
- Weight:0.84 lbs.
- Supplier no.:577-4983
- Handle type:L-Handle
Specifications
About this item
Proto® Long hex heat treated steel key sets.
- Rust-resistant black oxide finish
- Secure fastener fit
- Available in molded plastic holder
Delivery: Set includes 13 Hex keys per holder of different tip sizes 1/20", 1/16", 5/64", 3/32", 7/64", 1/8", 9/64", 5/32", 3/16", 7/32", 1/4", 5/16", 3/8".