Proto® Drift Punch Set, ORS Nasco
Catalog # 300003-226
Supplier: Safety & Industrial Supplies
Specifications
- Description:Drift Punch Set
- Material:Alloy steel
- Cat. No.:300003-226
- Includes:Kit pouch; 7 punches
- Finish:Black oxide
- Supplier no.:577-96A
- Handle type:Reverse Taper
- Tip type:Round
Specifications
About this item
Made from S2 steel and specially heat-treated for greater strength and durability, with a modified parabolic striking end to help reduce mushrooming and chipping.
- Reverse taper to help prevent binding
- Black oxide finish to help prevent rust corrosion; extending tool life and performance
- English measurements
- Packaged 7 per kit pouch
A polished identification panel on one of the flats is marked with the tool number and size in a black inlay. Set offers a wide range of sizes to handle a variety of jobs. Includes a pouch for convenient storage.