The new line of BVC fluid aspirator systems for cell culture provide the flexibility to accommodate decontamination protocol whether autoclave or disinfect in the bottle.
- Reliable VACUUBRAND vacuum pump used and proven in chemistry applications
- “G” Models have a Glass Bottle for use with in-bottle bleach decontamination
- Suitable for two users with optional extension and hand controller
- 0.2 Micron Bio-filter helps protect pump and lab atmosphere
- Professional Model have a contact-free level sensor
Basic Model:
The basic model is unpowered version of the BVC aspirator systems, suitable for use with in-house vacuum systems. A mechanical regulator provides a suitable vacuum level in the bottle for aspiration, and disconnects it from the supply when that level is reached- no power required. This reduces demand on the vacuum supply, increasing efficiency while reducing noise and impact on other applications.
Control Model:
The BVC control unit features a powerful, whisper-quiet Vacuubrand chemistry designed vacuum pump, thereby eliminating the need for central vacuum supply. The pump is turned on and off automatically on demand when using the hand controller. An easy to clean touch panel adjusts and indicated the vacuum level switch point.
Professional Model:
The top of the line BVC Professional model adds a liquid level sensor outside the bottle without contacting bottle contents. This lets you concentrate fully on your work without worrying about overflow. A special disinfection program allows the aspiration of disinfectant after the system is shut down to decontaminate the hand control.