Order Entry
United States
ContactUsLinkComponent
Organic Reference Standards, Idarubicin Hydrochloride, RS, USP
Organic Reference Standards, Idarubicin Hydrochloride, RS, USP
Catalog # 101096-756
Supplier:  USP
CAS Number:  
Organic Reference Standards, Idarubicin Hydrochloride, RS, USP
Catalog # 101096-756
Supplier:  USP
Supplier Number:  1335701
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • CAS No.:
    57852-57-0
  • Description:
    Idarubicin Hydrochloride
  • Size:
    50 mg
  • Formula:
    C₂₆H₂₈ClNO₉
  • Description:
    Idarubicin Hydrochloride
  • Cat. No.:
    101096-756
  • Supplier no.:
    1335701

Specifications

About this item